missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) pyrithione

Product Code. 15595946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15595946 1 g 1g
15505956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595946 Supplier Thermo Scientific Acros Supplier No. 457210010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14915-37-8 | C10H8CuN2O2S2 | 315.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14915-37-8
Molecular Formula C10H8CuN2O2S2
Molecular Weight (g/mol) 315.86

Specifications

Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 315.86
Percent Purity ≥97.5% (HPLC)
Infrared Spectrum Authentic
Chemical Name or Material Copper(II) pyrithione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.