missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) picolinate, 98%, Thermo Scientific™

Product Code. 15585946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585946 25 g 25g
15565946 1 g 1g
15575946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15585946 Supplier Thermo Scientific Acros Supplier No. 457200250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14050-01-2
Molecular Formula C12H8CuN2O4
Molecular Weight (g/mol) 307.75
InChI Key GNWVUSJJFONIKF-UHFFFAOYSA-L
Synonym copper ii picolinate, unii-96e596pypp, copper picolinate, copper, bis picolinato, copper picolinate anhydrous, copper, bis 2-pyridinecarboxylato-kappan1,kappao2, copper 2+ ion dipicolinate, copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2
PubChem CID 14963587
IUPAC Name copper;pyridine-2-carboxylate
SMILES C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]

Specifications

Quantity 25 g
Assay Percent Range 97.5% min. (Iodometry)
Formula Weight 307.75
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.02
Chemical Name or Material Copper(II) picolinate
compliance-icons compliance-icons
Product Identifier
  • Copper(II) picolinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.