missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) phthalocyanine

Product Code. 11365867
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365867 250 g 250g
11375867 10 g 10g
11355867 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365867 Supplier Thermo Scientific Alfa Aesar Supplier No. 043650.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 147-14-8 | C32H16CuN8

Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160; Pigment Blue 15

Specifications

Melting Point 350°C
Color Blue to Purple
Quantity 250 g
Beilstein 4121848
Merck Index 14,2520
Solubility Information It is insoluble in water.
Formula Weight 576.08
Physical Form Powder
Chemical Name or Material Copper (II) phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.