missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) perchlorate hexahydrate, Reagent Grade

Product Code. 11331758
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331758 250 g 250g
11341758 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331758 Supplier Thermo Scientific Alfa Aesar Supplier No. 011651.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-46-9 | Cl2CuH12O14 | 370.528 g/mol

Copper(II) perchlorate hexahydrate is a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-46-9
Molecular Formula Cl2CuH12O14
Molecular Weight (g/mol) 370.528
MDL Number MFCD00149666
InChI Key NHELIHXBJRANPL-UHFFFAOYSA-L
Synonym copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
PubChem CID 12846356
IUPAC Name copper;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]

Specifications

Melting Point 82°C
Physical Form Crystalline
Quantity 250 g
UN Number UN1481
Merck Index 14,2647
Solubility Information Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
Formula Weight 370.53 (262.44 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.505
Sensitivity Hygroscopic
Density 2.23 g/mL
Chemical Name or Material Copper(II) perchlorate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper (II) perchlorate hexahydrat
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Copper (II) perchlorate hexahydrat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.