missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) perchlorate hexahydrate, 98%

Product Code. 10522991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10522991 100 g Glass bottle 100g
10012730 5 g Glass bottle 5g
10117853 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10522991 Supplier Thermo Scientific Acros Supplier No. 201361000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-46-9 | Cl2CuO8·6H2O | 370.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-46-9
Molecular Formula Cl2CuO8·6H2O
Molecular Weight (g/mol) 370.52
MDL Number MFCD00149666
InChI Key NHELIHXBJRANPL-UHFFFAOYSA-L
Synonym copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
PubChem CID 12846356
IUPAC Name copper;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]

Specifications

Color Blue
pH 4 to 7 (5% soln.)
Physical Form Adhering Crystals
Quantity 100 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula Cu(ClO4)2·6H2O
Solubility Information Solubility in water: soluble in water
Formula Weight 370.52
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 2.22
Density 2.2200g/mL
Chemical Name or Material Copper(II) perchlorate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) perchlorate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.