missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) oxide, pure, wire form, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Color | Black |
| pH | 7 |
| Melting Point | 1326°C |
| Quantity | 100 g |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Copper(II) oxide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- H412-Harmful to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction