missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide, TRC

Product Code. 30137857
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137857 25 g 25g
30135099 50 g 50g
30011741 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137857 Supplier Toronto Research Chemicals Supplier No. TRCC68535025G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
Synonym BYK-LP x 20704, Banacobru OL, Black copper oxide, C.I. 77403, C.I. Pigment Black 15, Coopers Permatrace Copper, Copacaps, Copinox Lamb, Copper Brown, Copper monooxide, Copper monoxide, Copper monoxide (CuO), Copper oxide, Copper oxide (Cu4O4), Copper(2+) oxide, Copper(II) oxide, Copper(II) oxide (CuO), Copporal, Cupric oxide, ET, N 120, N 120 (oxide), N 520, NSC 83537, NanoActive CuO, NanoArc U 1102DBE, Nanotek CuO, Nanotek CuO-C, YC
IUPAC Name oxocopper
SMILES O=[Cu]

Specifications

Quantity 25 g
InChI Formula InChI=1S/Cu.O
Formula Weight 78.9245
Chemical Name or Material Copper(II) oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.