missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide, Technical, Powder, Fisher Chemical™

Product Code. p-7072945 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10111950

Brand: Fisher Chemical C/8200/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
IUPAC Name copper(2+) oxidandiide
SMILES [O--].[Cu++]

Specifications

Melting Point 1326°C
Color Black
pH 7
Physical Form Solid
Quantity 500 g
Formula Weight 79.54g/mol
Packaging Glass Jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.