missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Oxide, Certified AR for Analysis, Fisher Chemical

Product Code. 11473663 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473663 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473663 Supplier Fisher Chemical Supplier No. C/8360/50
  • ONLINE EXCLUSIVE PRICE
    £596.00
    £585.00 / 250g
    Save £11.00
    1% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point 1326°C
CAS 1317-38-0
Color Black
pH 7
Physical Form Solid
Quantity 250 g
Molecular Formula CuO
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES [O--].[Cu++]
IUPAC Name copper(2+) oxidandiide
Molecular Weight (g/mol) 79.55
Formula Weight 79.54g/mol
Packaging Plastic container
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.