missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, ACS reagent, powder

Product Code. 10288803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10288803 1 kg Plastic bottle 1kg
10528702 25 g Glass Bottle 25g
10137510 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10288803 Supplier Thermo Scientific Acros Supplier No. 405860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-38-0 | CuO | 79.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1326.0°C
Color Black
Physical Form Powder
Quantity 1 kg
Linear Formula CuO
Fieser 04,108; 05,161
Merck Index 15, 2636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dil.acids- slowly soluble in nh3,soluble in alkali cyanides and (nh4)2co3 solutions,practically insoluble in alcohol
IUPAC Name copper(2+) oxidandiide
Formula Weight 79.54
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Copper(II) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Copper(II) oxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.