missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99.9995% (metals basis)

Code produit 11448510
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
100 g
Conditionnement:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
11448510 25 g 25g
11438510 5 g 5g
11428510 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11448510 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 044356.14
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1317-38-0 | CuO | 79.55 g/mol

Copper(II) oxide is used as an adsorbent and absorbent, pigment, antifouling agent, wood preservative, optical glass polishing agent, welding flux for bronze materials, agricultural chemical and abrasive. It is used in the preparation of intermediates, oxidizing/reducing agents, surface treating agents and process regulators. It finds use in the manufacture of rayons, phosphors, and dry battery cells. It serves as a catalyst in certain chemical reactions of industrial importance. It is also employed in chemical sensors and superconducting materials. Hazardous materials like cyanides, dioxins and halogenated hydrocarbons can be disposed through oxidation with copper oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
IUPAC Name oxocopper
SMILES [O--].[Cu++]

Spécification

Melting Point 1326°C
Color Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9995% (Metal basis)
Solubility Information Practically insoluble in water,and alcohol. Soluble in dilute acids,alkali cyanides,and ammonium carbonate solution.
Formula Weight 79.55
Odor Odorless
Refractive Index 2.63
Appearance Black powder
Density 6.315 g/cm3
Chemical Name or Material Copper(II) oxide
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.