missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Oxide, 98%, Honeywell Fluka™

Product Code. 15691870 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
25g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691870 500 g Poly Bottle 500g
15671870 25 g Poly Bottle 25g
15681870 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691870 Supplier Honeywell-Fluka Supplier No. 208841500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, <10 μm, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1336°C
Color Black to Gray
Physical Form Powder
Quantity 500 g
UN Number UN3077
IUPAC Name copper(2+) oxidandiide
Formula Weight 79.55g/mol
Percent Purity 98%
Packaging Poly Bottle
Density 6.450g/cu.cm
Chemical Name or Material Copper(Ii) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.