missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 97%

Product Code. 10300694 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10300694

Brand: Thermo Scientific Acros 197440025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-38-0 | CuO | 79.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]

Specifications

Melting Point 1326.0°C
Color Black
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 96% min. (Iodometry)
Linear Formula CuO
Merck Index 15, 2636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dil.acids- slowly soluble in nh3,soluble in alkali cyanides and (nh4)2co3 solutions,practically insoluble in alcohol
IUPAC Name oxocopper
Formula Weight 79.54
Percent Purity 97%
Packaging Plastic bottle
Chemical Name or Material Copper(II) oxide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Copper(II) oxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.