missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Nitrate Hemipentahydrate, TRC

Product Code. 30357926
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357926 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357926 Supplier Toronto Research Chemicals Supplier No. TRCC685345500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) Nitrate Hemipentahydrate
CAS 19004-19-4
Molecular Formula Cu(NO3)2 • 2.5[H2O]
Quantity 500 g
Synonym Copper(II) Nitrate Hydrate (2:5)
InChI Formula InChI=1S/Cu.2NO3/c;2*2-1(3)4/q+2;2*-1
SMILES O=[N+](O[Cu]O[N+]([O-])=O)[O-]
IUPAC Name bis(nitrooxy)copper pentahydratehydrate
Molecular Weight (g/mol) 232.59
Formula Weight 231.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.