missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Nitrate Hemi(Pentahydrate), Honeywell Fluka™

Product Code. 15691200 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15691200 500 g 500g
15671200 1 kg 1kg
15681200 2.5 kg 2.5kg
15601210 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15691200 Supplier Honeywell-Fluka Supplier No. 12837500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

≥98%,≥26% Cu basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
MDL Number MFCD02253001
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Quantity 500 g
Linear Formula Cu(NO3)2 · 2.5H2O
UN Number 3085
Formula Weight 232.59g/mol
Percent Purity ≥98%
Packaging Plastic Bottle
Chemical Name or Material Copper(Ii) Nitrate Hemi(Pentahydrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.