missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Nitrate Hemi(Pentahydrate), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | Cu(NO3)2 · 2.5H2O |
| UN Number | 3085 |
| Formula Weight | 232.59g/mol |
| Percent Purity | ≥98% |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Copper(Ii) Nitrate Hemi(Pentahydrate) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction