missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) methoxide, typically 98% (metals basis)

Product Code. 11303507
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303507 5 g 5g
11393497 1 g 1g
11383497 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11303507 Supplier Thermo Scientific Alfa Aesar Supplier No. 018861.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1184-54-9

Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-54-9
MDL Number MFCD00015611

Specifications

Melting Point ∼206°C (decomposition)
Color Green
Odor Alcohol-like
Linear Formula Cu(OCH3)2
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 125.61
Percent Purity 98% (typically)
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Copper(II) methoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.