missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) methacrylate, tech., Thermo Scientific Chemicals

Catalog Number p-7051596
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
19662-59-0
C4H5CuO2
148.63
MFCD00080547,MFCD00156451,MFCD00080547
BFHDHRZKYIRNRP-UHFFFAOYSA-M
121233728
copper;2-methylprop-2-enoic acid;hydrate
[Cu++].CC(=C)C([O-])=O
This item is not returnable. View return policy
19662-59-0
C4H5CuO2
148.63
MFCD00080547,MFCD00156451,MFCD00080547
BFHDHRZKYIRNRP-UHFFFAOYSA-M
121233728
copper;2-methylprop-2-enoic acid;hydrate
[Cu++].CC(=C)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue-Green
Linear Formula [H2C=C(CH3)CO2]2Cu
Quantity 5 g
Formula Weight 233.71
Grade Technical
Physical Form Powder
Chemical Name or Material Copper(II) methacrylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'