missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) isopropoxide, 98% (metals basis)

Product Code. 11308727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308727 25 g 25g
11318727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308727 Supplier Thermo Scientific Alfa Aesar Supplier No. 035821.14
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23578-23-6 | C6H14CuO2 | 181.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23578-23-6
Molecular Formula C6H14CuO2
Molecular Weight (g/mol) 181.72
MDL Number MFCD00074982
InChI Key VNGORJHUDAPOQZ-UHFFFAOYSA-N
Synonym copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
PubChem CID 14389592
IUPAC Name copper;propan-2-olate
SMILES [Cu++].CC(C)[O-].CC(C)[O-]

Specifications

Quantity 25 g
UN Number UN3180
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 181.73
Percent Purity 98%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Copper(II) isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.