missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) hydroxide, tech. 94%, stab.

Product Code. p-3257000
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11362988

Brand: Thermo Scientific Alfa Aesar 032733.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used As a Raw Material in Chemical Processing and Catalyst Manufacturing | CAS: 20427-59-2 | H2CuO2

Used as a raw material in chemical processing and catalyst manufacturing. Copper(II) hydroxide is used as ceramic colorant. In organic synthesis, it is used as a catalyst to prepare 1-((2-aminoethyl)amino)anthraquinone, 1-amino-4-((2-aminoethyl)amino)anthraquinone by reacting with 1-bromoanthraquinone or 1-amino-4-bromoanthraquinone respectively. It is involved in the synthesis of benzoic acid and octanoic acid. It is used as a precursor in the production of rayon, battery electrodes and other copper salts It is also used as a dye mordant, paper colorant, pigment and feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20427-59-2
Molecular Formula H2CuO2
MDL Number MFCD00010968

Specifications

Color Blue
Physical Form Powder
Quantity 500 g
Assay Percent Range 94%
Solubility Information Slightly soluble in waterSoluble in acids,ammonium hydroxide,dilute hydrochloric acid,concentrated alkali and potassium cyanide. Insoluble in water,ethanol,and acetone.
Formula Weight 97.55
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Appearance Blue powder
Density 3.4 g/cm3
Chemical Name or Material Copper(II) hydroxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Copper hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.