missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) hydroxide, tech. 94%, stab.

Product Code. 11352988
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352988 2 kg 2kg
11362988 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352988 Supplier Thermo Scientific Alfa Aesar Supplier No. 032733.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used As a Raw Material in Chemical Processing and Catalyst Manufacturing | CAS: 20427-59-2 | H2CuO2

Used as a raw material in chemical processing and catalyst manufacturing. Copper(II) hydroxide is used as ceramic colorant. In organic synthesis, it is used as a catalyst to prepare 1-((2-aminoethyl)amino)anthraquinone, 1-amino-4-((2-aminoethyl)amino)anthraquinone by reacting with 1-bromoanthraquinone or 1-amino-4-bromoanthraquinone respectively. It is involved in the synthesis of benzoic acid and octanoic acid. It is used as a precursor in the production of rayon, battery electrodes and other copper salts It is also used as a dye mordant, paper colorant, pigment and feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20427-59-2
Molecular Formula H2CuO2
MDL Number MFCD00010968

Specifications

Color Blue
Physical Form Powder
Quantity 2 kg
Assay Percent Range 94%
Solubility Information Slightly soluble in waterSoluble in acids,ammonium hydroxide,dilute hydrochloric acid,concentrated alkali and potassium cyanide. Insoluble in water,ethanol,and acetone.
Formula Weight 97.55
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Appearance Blue powder
Density 3.4
Chemical Name or Material Copper(II) hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.