missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%

Product Code. 11480858
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480858 1 g 1g
11490858 5 g 5g
11400868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480858 Supplier Thermo Scientific Alfa Aesar Supplier No. B25284.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 155640-85-0 | C10H2CuF12O4 | 477.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155640-85-0
Molecular Formula C10H2CuF12O4
Molecular Weight (g/mol) 477.65
MDL Number MFCD00151019
InChI Key HZXGNBMOOYOYIS-PAMPIZDHSA-L
Synonym copper ii hexafluoroacetylacetonate hydrate, bis hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoro-2,4-pentanedionate hydrate, copper hexafluoroacetylacetonate hydrate, copper ii hexafluoroacetyl-acetonate hydrate, hexafluoroacetylacetonato copper ii hydrate, copper ii hexafluoroacetylacetonate hydrate, elec. gr., bis 1,1,1,5,5,5-hexafluoroacetylacetonato copper ii hydrate, copper ii 1,1,1,5,5,5-hexafluoro-2,4-pentanedionate hydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy cuprio oxy pent-3-en-2-one hydrate
PubChem CID 56845342
SMILES [Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 220°C
Melting Point ∼100°C
Quantity 1 g
IUPAC Name copper;(E)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
Formula Weight 477.64 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Copper(II) hexafluoro-2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.