missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride dihydrate

Product Code. p-7049162
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302178 50 g 50g
11312178 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302178 Supplier Thermo Scientific Alfa Aesar Supplier No. 013076.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Flux in casting gray iron | CAS: 13454-88-1 | CuF2H4O2 | 137.57 g/mol

Copper(II) fluoride can also be used to form fluorobenzene from benzene, hydrochloric acid and oxygen. Using a metal fluoride that can successfully oxidize with a methyl bond. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-88-1
Molecular Formula CuF2H4O2
Molecular Weight (g/mol) 137.57
MDL Number MFCD00010977
InChI Key CTRICEWDSISGPV-UHFFFAOYSA-L
Synonym cupric fluoride dihydrate, unii-3a38pc42e9, copper ii fluoride dihydrate, copper difluoride, dihydrate, cupric fluoride dihydrate mi, copper fluoride cuf2 , dihydrate, acmc-1bwnw, copper 2+ dihydrate difluoride, copper 2+ ion dihydrate difluoride
PubChem CID 24884278
IUPAC Name copper;difluoride;dihydrate
SMILES O.O.[F-].[F-].[Cu++]

Specifications

Melting Point >130°C (decomposition)
Physical Form Powder
Quantity 50 g
UN Number UN3288
Merck Index 14,2638
Solubility Information Slightly soluble in cold water, hydrolyzes to oxyfluoride in hot water
Formula Weight 137.57 (101.54 Anhydrous)
Odor Odorless
Density 2.934 g/mL
Chemical Name or Material Copper(II) fluoride dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.