missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride, anhydrous, 99.5%

Product Code. 11337157
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337157 50 g 50g
11347157 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337157 Supplier Thermo Scientific Alfa Aesar Supplier No. 011489.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent | CAS: 7789-19-7

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent.Copper(II) fluoride is used in the preparation of fluorobenzene by reacting with benzene. It is also used in galvanic cells, high-temperature fluorinating agent and fluxing iron castings. Further, it is used as an efficient catalyst in Chan-Lam-Evans coupling reaction to synthesis the diaryl- ether compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-19-7
MDL Number MFCD00010977

Specifications

Melting Point 950°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
UN Number UN3260
Merck Index 14,2638
Solubility Information Solubility in water at 20° ≈ 4;7g/100mL
Formula Weight 101.54
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 4.85 g/mL
Chemical Name or Material Copper(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.