Learn More
Copper(II) ethylacetoacetate, 97%, Thermo Scientific Chemicals
£31.50 - £802.00
Chemical Identifiers
CAS | 14284-06-1 |
---|---|
Molecular Formula | C12H18CuO6 |
Molecular Weight (g/mol) | 321.82 |
MDL Number | MFCD00017759 |
InChI Key | OXFZSJOUPRCPJO-UHFFFAOYSA-N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324238
|
Thermo Scientific Alfa Aesar
041489.04 |
2 g |
£31.50
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314238
|
Thermo Scientific Alfa Aesar
041489.09 |
10 g |
£112.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304238
|
Thermo Scientific Alfa Aesar
041489.18 |
50 g |
£282.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334238
|
Thermo Scientific Alfa Aesar
041489.30 |
250 g |
£802.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Copper(II) ethylacetoacetate it acts as a common cu(+2) precursor. It is also used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14284-06-1 | |
321.82 | |
OXFZSJOUPRCPJO-UHFFFAOYSA-N |
C12H18CuO6 | |
MFCD00017759 |
Specifications
14284-06-1 | |
Odorless | |
MFCD00017759 | |
OXFZSJOUPRCPJO-UHFFFAOYSA-N | |
321.81 | |
Powder |
192°C | |
C12H18CuO6 | |
Soluble in ethanol | |
321.82 | |
97% | |
Copper(II) ethylacetoacetate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 238-181-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only