missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) ethylacetoacetate, 97%

Product Code. 11334238
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334238 250 g 250g
11324238 2 g 2g
11314238 10 g 10g
11304238 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11334238 Supplier Thermo Scientific Alfa Aesar Supplier No. 041489.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14284-06-1 | C12H18CuO6 | 321.82 g/mol

Copper(II) ethylacetoacetate it acts as a common cu(+2) precursor. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14284-06-1
Molecular Formula C12H18CuO6
Molecular Weight (g/mol) 321.82
MDL Number MFCD00017759
InChI Key OXFZSJOUPRCPJO-UHFFFAOYSA-N

Specifications

Melting Point 192°C
Odor Odorless
Quantity 250 g
Solubility Information Soluble in ethanol
Formula Weight 321.81
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Copper(II) ethylacetoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.