missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) ethoxide, 98%

Product Code. 15432657
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432657 1 g 1g
15442657 5 g 5g
15452657 25 g 25g
15462657 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15432657 Supplier Thermo Scientific Alfa Aesar Supplier No. 046316.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2850-65-9 | C4H10CuO2 | 153.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2850-65-9
Molecular Formula C4H10CuO2
Molecular Weight (g/mol) 153.668
MDL Number MFCD00015650
InChI Key CRCKGIUJMFFISH-UHFFFAOYSA-N
Synonym copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, acmc-20aldp, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
PubChem CID 14419876
IUPAC Name copper;ethanolate
SMILES CC[O-].CC[O-].[Cu+2]

Specifications

Melting Point ∼120°C (decomposition)
Color Green
Linear Formula Cu(OC2H5)2
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 153.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Copper (II) ethoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.