missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11360959
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11360959

Brand: Thermo Scientific Alfa Aesar 010698.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol

Copper(II) chloride hydrate promotes the hydrolysis of acetonides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name dichlorocopper;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 100°C (decomposition)
Color Blue-Green
pH 3.0 to 3.8
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN2802
Merck Index 14,2633
Solubility Information Soluble in water.
Formula Weight 134.45 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.510 g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) chloride hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.