missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10509190 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
250g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10509190 5 kg Plastic keg 5kg
10070060 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10509190 Supplier Fisher Chemical Supplier No. C/7920/65

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
IUPAC Name copper(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 598°C
Color Blue-Green
pH 3.0 to 3.8
Physical Form Solid
Quantity 5 kg
Formula Weight 170.48g/mol
Packaging Plastic keg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.