missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
32.34 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
32.34 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
1 kg
Specifications
Melting Point | 598°C |
Color | Blue-Green |
pH | 3.0 to 3.8 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 170.48g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Copper(II) chloride dihydrate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Copper (II) chloridedihydrate, Cupric chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'