missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride dihydrate, ACS, 99+%, Thermo Scientific Chemicals

Product Code. 11311819
Click to view available options
Quantity:
100 g
500 g
2 kg

Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name copper(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 100°C (decomposition)
Physical Form Granules
Quantity 100 g
Assay Percent Range >99%
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 170.48 (134.45Anhydrous)
Grade ACS Reagent
Sensitivity Air sensitive; Hygroscopic
Density 2.51 g/mL
Chemical Name or Material Copper(II) chloride dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'