missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride dihydrate, 99+%, ACS reagent

Product Code. 15189263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15189263 2.5 kg Plastic Bottle 2.5kg
10093650 100 g Plastic Bottle 100g
10519653 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15189263 Supplier Thermo Scientific Acros Supplier No. 315280025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10125-13-0 | Cl2CuH4O2 | 170.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name dichlorocopper;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 100.0°C
Color Colorless
Physical Form Liquid
Quantity 2.5 kg
Assay Percent Range 99% min. (Iodometry)
Linear Formula CuCl2·2H2O
UN Number UN 2802
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152
Merck Index 15,2623
Solubility Information Solubility in water: 1150g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in acetone and ethylacetate, slightly soluble in ether
Formula Weight 170.48
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic Bottle
Chemical Name or Material Copper(II) Chloride Dihydrate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.