missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog Number 11371809
Click to view available options
Quantity:
50 g
250 g
1 kg
7447-39-4
CuCl2
MFCD00010972
7447-39-4
CuCl2
MFCD00010972

For Isomerization and Cracking Catalyst, Mordant in Dyeing and Printing Fabrics, in Electroplating Baths For Plating Cu On Al

Isomerization and cracking catalyst, mordant in dyeing and printing fabrics, in electroplating baths for plating Cu on Al, in photography as a fixer, as a pigment for glass and ceramic.Copper(II) chloride is used as a co-catalyst with palladium(II) chloride used in the Wacker process to prepare ethanol from ethane. It acts as an effective catalyst for the tetrahydropyranylation of alcohols. It is involved in the production of vinyl chloride and dichloroethane as well as in the chlorination of aromatic hydrocarbons. It acts as deodorizing and desulfurizing agent in the petroleum industry, a mordant in textiles industry and in electrotyping baths for plating copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 993°C (decomposition)
Melting Point 620°C
Color Brown-Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,alcohol,acetone and hot sulfuric acid.
Formula Weight 134.45
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow-brown powder
Density 3.386 g/cm3
Chemical Name or Material Copper(II) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'