missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Carbonate, Technical, Basic, Fisher Chemical™

Product Code. 11443703 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
3 kg
Unit Size:
3kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443703 3 kg 3kg
11423723 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443703 Supplier Fisher Chemical Supplier No. C/7879/63
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Copper(II) Carbonate, Technical, Basic

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CH2Cu2O5
Molecular Weight (g/mol) 221.11
MDL Number 10976
InChI Key ZMMDPCMYTCRWFF-UHFFFAOYSA-J
IUPAC Name dicopper(2+) dihydroxide carbonate
SMILES [OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O

Specifications

Melting Point 200°C
Color Green
pH 8 to 9
Physical Form Powder/Solid
Quantity 3 kg
Formula Weight 221.1g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Copper (II) carbonate basic
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.