missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Carbonate Dihydroxide, TRC

Product Code. 30359221
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30359221

Brand: Toronto Research Chemicals TRCC6852855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) Carbonate Dihydroxide
CAS 12069-69-1
Molecular Formula CH2Cu2O5
Quantity 5 g
Synonym [μ-[Carbonato(2-)-kO:kO']]dihydroxydicopper, Copper Carbonate Hydroxide, (Carbonato)dihydroxydicopper, [μ-[Carbonato(2-)-O:O']]dihydroxydicopper, Carbonic Acid Copper Complex, Basic Copper Carbonate, Basic Copper Carbonate, Basic Copper Carbonate, Basic Copper(II) Carbonate, Basic Cupric Carbonate, Carbonatodihydroxodicopper, Basic Carbonic Acid Copper(2+) Salt (1:1), Cheshunt Compound, Copper Basic Carbonate, Copper Carbonate Hydroxide, Copper Dihydroxide Carbonate, Copper Hydroxide Carbonate, Cupric Carbonate, Cupric Carbonate Hydroxide, Basic Cupric Carbonate, Dicopper Dihydroxycarbonate, Dicopper(2+) Carbonate Dihydroxide
InChI Formula InChI=1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4
SMILES [Cu+2].O[Cu]O.[O-]C(=O)[O-]
IUPAC Name copper;dihydroxycopper;carbonate
Molecular Weight (g/mol) 221.12
Formula Weight 219.8494
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.