missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Carbonate Dihydroxide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Copper(II) Carbonate Dihydroxide |
| CAS | 12069-69-1 |
| Molecular Formula | CH2Cu2O5 |
| Quantity | 25 g |
| Synonym | [μ-[Carbonato(2-)-kO:kO']]dihydroxydicopper, Copper Carbonate Hydroxide, (Carbonato)dihydroxydicopper, [μ-[Carbonato(2-)-O:O']]dihydroxydicopper, Carbonic Acid Copper Complex, Basic Copper Carbonate, Basic Copper Carbonate, Basic Copper Carbonate, Basic Copper(II) Carbonate, Basic Cupric Carbonate, Carbonatodihydroxodicopper, Basic Carbonic Acid Copper(2+) Salt (1:1), Cheshunt Compound, Copper Basic Carbonate, Copper Carbonate Hydroxide, Copper Dihydroxide Carbonate, Copper Hydroxide Carbonate, Cupric Carbonate, Cupric Carbonate Hydroxide, Basic Cupric Carbonate, Dicopper Dihydroxycarbonate, Dicopper(2+) Carbonate Dihydroxide |
| InChI Formula | InChI=1S/CH2O3.2Cu.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4 |
| SMILES | [Cu+2].O[Cu]O.[O-]C(=O)[O-] |
| IUPAC Name | copper;dihydroxycopper;carbonate |
| Molecular Weight (g/mol) | 221.12 |
| Formula Weight | 219.8494 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction