missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) carbonate basic, Puriss., 54-57% Cu basis, green, Honeywell Fluka™

Product Code. 15651200 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15651200 250 g 250g
15641200 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15651200 Supplier Honeywell-Fluka Supplier No. 12830250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., 54-57% Cu basis, green

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Synonym Cupric carbonate basic

Specifications

Quantity 250 g
Assay Percent Range 54 to 57%
Linear Formula C2H5OCH2CH2OH
MDL Number MFCD00002869
Formula Weight 221.12
Grade puriss.
Packaging Poly Bottle
Density 4 g/cm3
Chemical Name or Material Copper(II) carbonate basic, Puriss., 54-57% Cu basis, green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.