missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) carbonate basic, Solstice

Product Code. 15616020
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15616020 1 kg 1kg
15626020 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15616020 Supplier Fluka Supplier No. 611671KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Purum p.a.,≥95% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
MDL Number MFCD00010976

Specifications

Quantity 1 kg
Linear Formula CuCO3 · Cu(OH)2
UN Number NONH for all modes of transport
Formula Weight 221.12g/mol
Percent Purity ≥95% (RT)
Chemical Name or Material Copper(II) carbonate basic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.