missing translation for 'onlineSavingsMsg'
Learn More

Copper (II) Bromide, TRC

Product Code. 30351163
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30351163 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351163 Supplier Toronto Research Chemicals Supplier No. TRCC68527550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) bromide
CAS 7789-45-9
Molecular Formula Br2 Cu
Quantity 50 g
Synonym Copper Dibromide, Copper(II) Bromide, Cupric Bromide
InChI Formula InChI=1S/2BrH.Cu/h2*1H;/q;;+2/p-2
SMILES Br[Cu]Br
IUPAC Name dibromocopper
Molecular Weight (g/mol) 223.35
Formula Weight 220.7663
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.