missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Bromide, Technical, SLR, Fisher Chemical

Product Code. 10578990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10578990 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10578990 Supplier Fisher Chemical Supplier No. C/7840/50
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 900°C
Melting Point 498°C
CAS 7789-45-9
Color Gray
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula Br2Cu
MDL Number 10970
InChI Key QTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES [Cu++].[Br-].[Br-]
IUPAC Name copper(2+) dibromide
Molecular Weight (g/mol) 223.35
Formula Weight 223.36g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.