missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 99+%, extra pure, anhydrous

Product Code. 10422401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422401 5 g Glass bottle 5g
10144880 100 g Glass bottle 100g
10398653 500 g Glass bottle 500g
10493065 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10422401 Supplier Thermo Scientific Acros Supplier No. 206330050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-45-9 | Br2Cu | 223.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-45-9
Molecular Formula Br2Cu
Molecular Weight (g/mol) 223.36
MDL Number MFCD00010970
Synonym Cupric bromide

Specifications

Boiling Point 900.0°C
Melting Point 498.0°C
Color Black-Gray to Gray
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula CuBr2
Fieser 01,161; 02,84; 05,158; 06,138; 13,112; 15,100
Merck Index 15, 2619
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, acetone, nh3, practically insoluble in benzene, diethyl ether,, conc. h2so4
Formula Weight 223.36
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Density 4.77
Chemical Name or Material Copper(II) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.