missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) bromide, 98+%

Product Code. 11397837
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397837 500 g 500g
11387837 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397837 Supplier Thermo Scientific Alfa Aesar Supplier No. 014258.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-45-9 | CuBr2

Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-45-9
Molecular Formula CuBr2
MDL Number MFCD00010970

Specifications

Boiling Point 900°C
Melting Point 498°C
Color Black
Physical Form Crystalline
Quantity 500 g
Solubility Information Soluble in water,alcohol,ammonia and acetone. Insoluble in benzene,ether,sulfuric acid and ethyl ether.
Formula Weight 223.36
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Appearance Black Crystalline powder
Density 4.71
Chemical Name or Material Copper(II) bromide, Cu 28.1% min.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.