missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, Solstice

Product Code. 15656010 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15656010 100 g 100g
15666010 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15656010 Supplier Honeywell-Fluka Supplier No. 61148100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Quantity 100 g
Linear Formula Cu(CO2CH3)2 · H2O
UN Number UN1759
Beilstein 3730548
IUPAC Name copper;diacetate;hydrate
Formula Weight 199.65g/mol
Percent Purity ≥99% (RT)
Density Vapor Density: 6.8 (vs air)
Chemical Name or Material Copper(II) acetate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.