missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, ACS, 98.0-102.0%

Product Code. 11337767
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337767 500 g 500g
11347767 100 g 100g
11357767 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337767 Supplier Thermo Scientific Alfa Aesar Supplier No. 014177.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6046-93-1
Molecular Formula C4H8CuO5
Molecular Weight (g/mol) 199.65
MDL Number MFCD00149570
InChI Key NWFNSTOSIVLCJA-UHFFFAOYSA-L
Synonym copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
PubChem CID 165397
IUPAC Name copper;diacetate;hydrate
SMILES O.[Cu++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 115°C
Color Blue-Green
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and alcohol. Slightly soluble in ether and glycerin.
Formula Weight 199.65 (181.64 Anhydrous)
Grade ACS
Density 1.882
Chemical Name or Material Copper(II) acetate, monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.