missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, 98+%, extra pure, Thermo Scientific Chemicals

Catalog Number 10328390
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Boiling Point 240.0°C
Melting Point 115.0°C
Color Blue to Blue-Green
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula (CH3CO2)2Cu·H2O
Merck Index 15,2614
Solubility Information Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 199.65
Percent Purity ≥98%
Grade Extra Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Density 1.88g/mL
Chemical Name or Material Copper(II) acetate monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) acetate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'