missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Acetate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10568800 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10568800 250 g 250g
10223520 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10568800 Supplier Fisher Chemical Supplier No. C/7760/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 142-71-2
Molecular Formula C4H6CuO4
Molecular Weight (g/mol) 181.63
MDL Number 8690
InChI Key OPQARKPSCNTWTJ-UHFFFAOYSA-L
IUPAC Name copper(2+) diacetate
SMILES [Cu++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 115°C
Color Blue-Green
Physical Form Solid
Quantity 250 g
Formula Weight 181.64g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) acetate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.