missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) 2-ethylhexanoate

Product Code. 11397947
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11397947

Brand: Thermo Scientific Alfa Aesar 020539.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-11-1 | C16H32CuO4 | 351.974 g/mol

Copper(II) 2-ethylhexanoate is used as pharmaceutical intermediates. Used as drier/additive for paints, coatings and inks. Used as catalystic special applications in lubricant and grease additives, fuel combustion additives to enhance and stabilize the performance of fuels. Used in the the rubber wire coat compound and the brass-coated steel belt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-11-1
Molecular Formula C16H32CuO4
Molecular Weight (g/mol) 351.974
MDL Number MFCD00015695
InChI Key SZXCZUDYSKBGSX-UHFFFAOYSA-N
Synonym copper bis 2-ethylhexanoate
PubChem CID 85309523
IUPAC Name copper;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Cu]

Specifications

Melting Point 252°C (decomposition)
Color Blue-Green
Quantity 50 g
Solubility Information Soluble in water,alcohols,dichloroethane,chlorobenzene,and most organic solvents.
Formula Weight 349.96
Physical Form Powder or Lumps
Chemical Name or Material Copper(II) 2-ethylhexanoate
compliance-icons
Product Identifier
  • Copper(II) 2-ethylhexanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Copper(II) 2-ethylhexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.