missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) trifluoromethanesulfonate benzene complex, 90%

Product Code. 10028971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10028971 1 g Glass bottle 1g
10293052 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10028971 Supplier Thermo Scientific Acros Supplier No. 371090010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42152-46-5 | C8H6Cu2F6O6S2 | 503.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42152-46-5
Molecular Formula C8H6Cu2F6O6S2
Molecular Weight (g/mol) 503.33
MDL Number MFCD00043300
Synonym Trifluoromethanesulfonic acid, copper(I) salt, benzene complex

Specifications

Melting Point 160.0°C
Color Gray-Green
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula (CF3SO3Cu)2C6H6
Quantity 1 g
Fieser 12,144; 16,99
Solubility Information Solubility in water: .. Other solubilities: soluble in benzene,toluene
Formula Weight 503.33
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Copper(I) trifluoromethanesulfonate benzene complex
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(I) trifluoromethanesulfonate benzene complex
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.