missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) thiocyanate, 96% min

Product Code. 11329377
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329377 50 g 50g
11339377 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329377 Supplier Thermo Scientific Alfa Aesar Supplier No. 040220.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1111-67-7 | CHCuNS | 122.632 g/mol

It is used as flame retardant. It is a good inorganic pigment, and is used as the antifouling paint for protecting the underwater surfaces of ships against vegetation. Manufacturing industrial chemicals, pharmaceuticals and also used in freezing solutions, electroplating , steel picking, printing, and corrosion inhibitor against acid gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-67-7
Molecular Formula CHCuNS
Molecular Weight (g/mol) 122.632
MDL Number MFCD00010980
InChI Key WEKJRGBVEBCTRF-UHFFFAOYSA-N
Synonym copper i thio-cyanate, thiocyanate ion copper
PubChem CID 16211891
IUPAC Name copper;thiocyanic acid
SMILES C(#N)S.[Cu]

Specifications

Melting Point 1084°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 96% min.
Solubility Information Practically insoluble in water,alcohol. Soluble in NH4OH,ether
Formula Weight 121.63
Odor Odorless
Density 2.84 g/cm3
Chemical Name or Material Copper(I) thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.