missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) telluride, 99.5% (metals basis)

Product Code. 11362238
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11362238

Brand: Thermo Scientific Alfa Aesar 013140.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12019-52-2 | Cu2Te | 254.692 g/mol

It is used as a colorent of glass and ceramics, rubber vulcanization accelerator, metallurgical additives, petrochemical catalyst, solar cells, thermoelectric refrigeration Materials, Infrared Detection Materials, Electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12019-52-2
Molecular Formula Cu2Te
Molecular Weight (g/mol) 254.692
MDL Number MFCD00049727
InChI Key MZEWONGNQNXVKA-UHFFFAOYSA-N
Synonym copper telluride, cuprous telluride, copper i telluride, dicopper telluride, cuprous monotelluride, dicopper monotelluride, copper telluride cu0.67te0.33, cupriotellanyl copper
PubChem CID 6914517
IUPAC Name copper;tellurium
SMILES [Cu].[Cu].[Te]

Specifications

Color Black-Blue
Physical Form Pieces
Quantity 25 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 254.68
Odor Odorless
Refractive Index 1.45
Density 7.27
Chemical Name or Material Copper(I) telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.